Ytterbium(III) oxide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Y0008-25G Y0008-5G
TCY0008-25GEA 154.84 CAD
TCY0008-25G TCY0008-5G
Ytterbium(III) oxide ≥98.0% (by titrimetric analysis)
Ytterbium (III) oxide

CAS Number: 1314-37-0
MDL Number: MFCD00011290
Molecular Formula: O3Yb2
Molecular Weight: 394.11
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White

Formula: O₃Yb₂
MDL Number: MFCD00011290
CAS Number: 1314-37-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ffdd20403a6dfb8c94b600a38cb4fe3e72f8e53e&vid=ffdd20403a6dfb8c94b600a38cb4fe3e72f8e53e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR