(S)-(-)-XylBINAP ≥97.0% (by HPLC)

Supplier: TCI America

Synonyms: (S)-2,2'-Bis(bis(3,5-dimethylphenyl)phosphino)-1,1'-binaphthalene, (S)-(-)-2,2'-Bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl

X0071-1G X0071-200MG
TCX0071-1GEA 307.47 CAD
TCX0071-1G TCX0071-200MG
(S)-(-)-XylBINAP ≥97.0% (by HPLC)
(S)-(-)-XylBINAP

CAS Number: 135139-00-3
MDL Number: MFCD01630821
Molecular Formula: C52H48P2
Molecular Weight: 734.90
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 255
Specific rotation [a]20/D: -220.8 deg (C=1, CHCl3)


Product Categories: Synthetic Organic Chemistry, Asymmetric Synthesis, Ligands, Ligands ([2+2+2]Cycloaddition Reaction)
Other Product Names / Synonyms: (S)-(-)-2,2'-Bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl

Formula: C₅₂H₄₈P₂
MW: 734.90 g/mol
Melting Pt: 255 °C
MDL Number: MFCD01630821
CAS Number: 135139-00-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d0b1fc5ffe0e1257f25ea0522d25f3cb7f78a7df&vid=d0b1fc5ffe0e1257f25ea0522d25f3cb7f78a7df&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR