(R)-(+)-XylBINAP ≥97.0% (by HPLC)

Supplier: TCI America

X0070-1G X0070-200MG
TCX0070-1GEA 302.41 CAD
TCX0070-1G TCX0070-200MG
(R)-(+)-XylBINAP ≥97.0% (by HPLC)
(R)-(+)-XylBINAP

CAS Number: 137219-86-4
MDL Number: MFCD01630821
Molecular Formula: C52H48P2
Molecular Weight: 734.90
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 191
Specific rotation [a]20/D: 169 deg (C=1, CHCl3)


Product Categories: Synthetic Organic Chemistry, Asymmetric Synthesis, Ligands, Ligands ([2+2+2]Cycloaddition Reaction)
Other Product Names / Synonyms: (R)-(+)-2,2'-Bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl

Formula: C₅₂H₄₈P₂
Melting Pt: 191 °C
MDL Number: MFCD01630821
CAS Number: 137219-86-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c3d8048a545dbf8e1bcda0d47e5736c9f9ba3212&vid=c3d8048a545dbf8e1bcda0d47e5736c9f9ba3212&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR