vBRIDP® ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: Di-tert-butyl(1,1-diphenyl-1-propen-2-yl)phosphine

V0132-1G V0132-200MG
TCV0132-1GEA 198.29 CAD
TCV0132-1G TCV0132-200MG
vBRIDP® ≥98.0% (by HPLC)
vBRIDP®

CAS Number: 384842-25-5
MDL Number: MFCD16621412
Molecular Formula: C23H31P
Molecular Weight: 338.47
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 133


Other Product Names / Synonyms: Di-tert-butyl(1,1-diphenyl-1-propen-2-yl)phosphine

Formula: C₂₃H₃₁P
MW: 338.47 g/mol
Melting Pt: 133 °C
MDL Number: MFCD16621412
CAS Number: 384842-25-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=74ad6655fa316ae072fc5cf9ec7bcaf6e718ca48&vid=74ad6655fa316ae072fc5cf9ec7bcaf6e718ca48&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR