DL-Valine methyl ester hydrochloride ≥98.0%

Supplier: TCI America

Synonyms: H-DL-Val-OMe.HCl, H-DL-Val-OMe·HCl

V0125-25G V0125-5G
TCV0125-25GEA 152 CAD
TCV0125-25G TCV0125-5G
DL-Valine methyl ester hydrochloride ≥98.0%
DL-Valine methyl ester hydrochloride

CAS Number: 5619-05-6
MDL Number: MFCD00066118
Molecular Formula: C6H13NO2
Molecular Weight: 167.63
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 105


Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: H-DL-Val-OMe.HCl

Formula: C₆H₁₃NO₂
Melting Pt: 105 °C
MDL Number: MFCD00066118
CAS Number: 5619-05-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0bcffb8ca04f21b0fe3307322176f761c5e878c0&vid=0bcffb8ca04f21b0fe3307322176f761c5e878c0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR