Vinpocetine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
(41S,13aS)-Ethyl 13a-ethyl-2,3,41,5,6,13a-hexahydro-1H-indolo[3,2,1-de]pyrido[3,2,1-ij][1,5]naphthyridine-12-carboxylate
V0115-100MG
V0115-1G
TCV0115-100MGEA
135.56
CAD
TCV0115-100MG
TCV0115-1G
Vinpocetine ≥98.0% (by HPLC, titration analysis)
Vinpocetine
CAS Number: 42971-09-5
MDL Number: MFCD00211233
Molecular Formula: C22H26N2O2
Molecular Weight: 350.46
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 151
Specific rotation [a]20/D: 118 deg (C=2, Pyridine)
Lambda max.: 315 nm (EtOH)
Formula:
C₂₂H₂₆N₂O₂ MW: 350.46 g/mol Melting Pt: 151 °C |
MDL Number:
MFCD00211233 CAS Number: 42971-09-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...