Undec-10-enoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

U0007-25G U0007-400G
TCU0007-25GEA 28.44 CAD
TCU0007-25G TCU0007-400G
Undec-10-enoic acid ≥98.0% (by GC, titration analysis)
Undec-10-enoic acid

CAS Number: 112-38-9
MDL Number: MFCD00004442
Molecular Formula: C11H20O2
Molecular Weight: 184.28
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 275
Flash Point (°C): 146
Freezing point (°C): 24


Other Product Names / Synonyms: Undecylenic Acid

Formula: C₁₁H₂₀O₂
Boiling Pt: 275 °C
Flash Pt: 146 °C
MDL Number: MFCD00004442
CAS Number: 112-38-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9fa48934975d805ed34ebff7017bce7af747a920&vid=9fa48934975d805ed34ebff7017bce7af747a920&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR