1,3,5-Tris(3,5-dibromophenyl)benzene ≥95.0% (by HPLC)

Supplier: TCI America

T3213-1G T3213-200MG
TCT3213-1GEA 816.54 CAD
TCT3213-1G TCT3213-200MG
1,3,5-Tris(3,5-dibromophenyl)benzene ≥95.0% (by HPLC)
1,3,5-Tris(3,5-dibromophenyl)benzene

CAS Number: 29102-67-8
MDL Number: MFCD13193235
Molecular Formula: C24H12Br6
Molecular Weight: 779.78
Purity/Analysis Method: >95.0% (HPLC)
Form: Crystal
Color: Pale Yellow
Melting point (°C): 315

Formula: C₂₄H₁₂Br₆
Melting Pt: 315 °C
MDL Number: MFCD13193235
CAS Number: 29102-67-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fa3f7bf0a9fbbadad4cdecd98d6a140348228034&vid=fa3f7bf0a9fbbadad4cdecd98d6a140348228034&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR