Thieno[3,2-b]thiophene-2,5-dicarboxaldehyde ≥93.0%
Supplier: TCI America
Synonyms:
Thieno[3,2-b]thiophene-2,5-dicarbaldehyde
T3212-1G
T3212-200MG
TCT3212-1GEA
469.26
CAD
TCT3212-1G
TCT3212-200MG
Thieno[3,2-b]thiophene-2,5-dicarboxaldehyde ≥93.0%
Thieno[3,2-b]thiophene-2,5-dicarboxaldehyde
CAS Number: 37882-75-0
MDL Number: MFCD03931117
Molecular Formula: C8H4O2S2
Molecular Weight: 196.24
Purity/Analysis Method: >93.0% (GC)
Form: Crystal
Color: Reddish Yellow
Melting point (°C): 274
Formula:
C₈H₄O₂S₂ MW: 196.24 g/mol Melting Pt: 274 °C |
MDL Number:
MFCD03931117 CAS Number: 37882-75-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...