3-(Trimethylsilylethynyl)thiophene ≥98.0% (by GC)
Supplier: TCI America
T3190-1G
T3190-5G
TCT3190-1GEA
58.46
CAD
TCT3190-1G
TCT3190-5G
3-(Trimethylsilylethynyl)thiophene ≥98.0% (by GC)
3-(Trimethylsilylethynyl)thiophene
CAS Number: 130995-13-0
MDL Number: MFCD04039959
Molecular Formula: C9H12SSi
Molecular Weight: 180.34
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Slightly Pale Yellow
Flash Point (°C): 89
Specific Gravity (20/20): 0.98
Formula:
C₉H₁₂SSi Density: 0.98 (20 °C) Flash Pt: 89 °C |
MDL Number:
MFCD04039959 CAS Number: 130995-13-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...