Trimethoxy(4-vinylphenyl)silane ≥97.0% (by GC)

Supplier: TCI America

T3185-25G T3185-5G
TCT3185-25GEA 636.74 CAD
TCT3185-25G TCT3185-5G
Trimethoxy(4-vinylphenyl)silane ≥97.0% (by GC)
Trimethoxy(4-vinylphenyl)silane

CAS Number: 18001-13-3
MDL Number: MFCD21596662
Molecular Formula: C11H16O3Si
Molecular Weight: 224.33
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 77
Flash Point (°C): 94
Specific Gravity (20/20): 1.06

Formula: C₁₁H₁₆O₃Si
Boiling Pt: 77 °C
Density: 1.06 (20 °C)
Flash Pt: 94 °C
MDL Number: MFCD21596662
CAS Number: 18001-13-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d2fefa74a2e8baa1f6b10c9849004ef769fa954e&vid=d2fefa74a2e8baa1f6b10c9849004ef769fa954e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR