Triethylene glycol butyl methyl ether ≥98.0%

Supplier: TCI America

Synonyms: 1-[2-[2-(2-Methoxyethoxy)ethoxy]ethoxy]butane

T3181-25ML T3181-500ML
TCT3181-25MLEA 33.18 CAD
TCT3181-25ML TCT3181-500ML
Triethylene glycol butyl methyl ether ≥98.0%
Triethylene glycol butyl methyl ether

CAS Number: 7382-30-1
MDL Number: MFCD23703159
Molecular Formula: C11H24O4
Molecular Weight: 220.31
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Melting point (°C): -34
Specific Gravity (20/20): 0.94


Other Product Names / Synonyms: 1-[2-[2-(2-Methoxyethoxy)ethoxy]ethoxy]butane

Formula: C₁₁H₂₄O₄
MW: 220.31 g/mol
Melting Pt: –34 °C
Density: 0.94 (20 °C)
MDL Number: MFCD23703159
CAS Number: 7382-30-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea123c9a3c52ce640d589b8aac0ce2c3e294ed17&vid=ea123c9a3c52ce640d589b8aac0ce2c3e294ed17&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR