1-(Trifluoromethanesulfonyl)imidazole ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1-((Trifluoromethyl)sulphonyl)-1H-imidazole, 1-((Trifluoromethyl)sulfonyl)-1H-imidazole

T3177-1G T3177-5G
TCT3177-1GEA 381.89 CAD
TCT3177-1G TCT3177-5G
1-(Trifluoromethanesulfonyl)imidazole ≥98.0% (by GC)
1-(Trifluoromethanesulfonyl)imidazole

CAS Number: 29540-81-6
MDL Number: MFCD00037891
Molecular Formula: C4H3F3N2O2S
Molecular Weight: 200.14
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 40
Melting point (°C): 19
Specific Gravity (20/20): 1.53
Storage Temperature: 0-10°C

Formula: C₄H₃F₃N₂O₂S
MW: 200.14 g/mol
Boiling Pt: 40 °C
Melting Pt: 19 °C
Density: 1.53 (20 °C)
MDL Number: MFCD00037891
CAS Number: 29540-81-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4239625f996bee3e4e2bbbf40344f12332131261&vid=4239625f996bee3e4e2bbbf40344f12332131261&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR