Thieno[3,2-b]thiophene-2-carboxylic acid ≥98.0%
Supplier: TCI America
T3156-1G
T3156-200MG
TCT3156-1GEA
325.48
CAD
TCT3156-1G
TCT3156-200MG
Thieno[3,2-b]thiophene-2-carboxylic acid ≥98.0%
Thieno[3,2-b]thiophene-2-carboxylic acid
CAS Number: 1723-27-9
MDL Number: MFCD00115174
Molecular Formula: C7H4O2S2
Molecular Weight: 184.23
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 223
Formula:
C₇H₄O₂S₂ MW: 184.23 g/mol Melting Pt: 223 °C |
MDL Number:
MFCD00115174 CAS Number: 1723-27-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...