(S)-(-)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl ≥98.0% (by HPLC)
Supplier: TCI America
T3153-1G
T3153-5G
TCT3153-1GEA
122.29
CAD
TCT3153-1G
TCT3153-5G
(S)-(-)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl ≥98.0% (by HPLC)
(S)-(-)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl
CAS Number: 100165-88-6
MDL Number: MFCD01311709
Molecular Formula: C48H40P2
Molecular Weight: 678.80
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 258
Product Categories: Synthetic Organic Chemistry, Asymmetric Synthesis, Ligands, Ligands ([2+2+2]Cycloaddition Reaction)
Other Product Names / Synonyms: (S)-(-)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl
Formula:
C₄₈H₄₀P₂ Melting Pt: 258 °C |
MDL Number:
MFCD01311709 CAS Number: 100165-88-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...