Tetrakis(4-ethynylphenyl)methane ≥98.0% (by GC)

Supplier: TCI America

T3151-100MG T3151-1G
TCT3151-100MGEA 292.3 CAD
TCT3151-100MG TCT3151-1G
Tetrakis(4-ethynylphenyl)methane ≥98.0% (by GC)
Tetrakis(4-ethynylphenyl)methane

CAS Number: 177991-01-4
Molecular Formula: C33H20
Molecular Weight: 416.52
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: Slightly Pale Yellow
Melting point (°C): 300

Formula: C₃₃H₂₀
Melting Pt: 300 °C
MDL Number: MFCD24873189
CAS Number: 177991-01-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1054f6278fbded420864d03f0fbe469e5b49358c&vid=1054f6278fbded420864d03f0fbe469e5b49358c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR