3,3',5,5'-Tetramethylbiphenyl-4,4'-diol ≥98.0% (by GC)
Supplier: TCI America
T3145-5G
T3145-25G
TCT3145-5GEA
160.37
CAD
TCT3145-5G
TCT3145-25G
3,3',5,5'-Tetramethylbiphenyl-4,4'-diol ≥98.0% (by GC)
3,3',5,5'-Tetramethylbiphenyl-4,4'-diol
CAS Number: 2417-04-1
MDL Number: MFCD00094737
Molecular Formula: C16H18O2
Molecular Weight: 242.32
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 225
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 4,4'-Dihydroxy-3,3',5,5'-tetramethylbiphenyl
Formula:
C₁₆H₁₈O₂ Melting Pt: 225 °C |
MDL Number:
MFCD00094737 CAS Number: 2417-04-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...