2,4,6-Tribromophenyl acrylate ≥98.0% (by GC)

Supplier: TCI America

T3130-25G T3130-5G
TCT3130-25GEA 380.15 CAD
TCT3130-25G TCT3130-5G
2,4,6-Tribromophenyl acrylate ≥98.0% (by GC)
2,4,6-Tribromophenyl acrylate

CAS Number: 3741-77-3
MDL Number: MFCD00078430
Molecular Formula: C9H5Br3O2
Molecular Weight: 384.85
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 78


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Acrylic Acid 2,4,6-Tribromophenyl Ester

Formula: C₉H₅Br₃O₂
Melting Pt: 78 °C
MDL Number: MFCD00078430
CAS Number: 3741-77-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0d031fa067c5cf6ad2363bb54f1654fcff155486&vid=0d031fa067c5cf6ad2363bb54f1654fcff155486&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR