ɑ-(Trichloromethyl)benzyl acetate ≥98.0% (by GC)

Supplier: TCI America

T3122-250G T3122-25G
TCT3122-250GEA 123.24 CAD
TCT3122-250G TCT3122-25G
ɑ-(Trichloromethyl)benzyl acetate ≥98.0% (by GC)
alpha-(Trichloromethyl)benzyl acetate

CAS Number: 90-17-5
MDL Number: MFCD00000804
Molecular Formula: C10H9Cl3O2
Molecular Weight: 267.53
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 282
Melting point (°C): 88


Other Product Names / Synonyms: Acetic Acid alpha-(Trichloromethyl)benzyl Ester, 2,2,2-Trichloro-1-phenylethyl Acetate, Acetic Acid 2,2,2-Trichloro-1-phenylethyl Ester

Formula: C₁₀H₉Cl₃O₂
Boiling Pt: 282 °C
Melting Pt: 88 °C
MDL Number: MFCD00000804
CAS Number: 90-17-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c831d24dfa21dc39a51183d9faa519050ce5c32d&vid=c831d24dfa21dc39a51183d9faa519050ce5c32d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR