2,4,6-Trichlorophenyl formate ≥97.0% (by HPLC)

Supplier: TCI America

Synonyms: Formic Acid 2,4,6-Trichlorophenyl Ester, 2,4,6-Trichlorophenyl Formate

T3121-1G T3121-5G
TCT3121-1GEA 53.72 CAD
TCT3121-1G TCT3121-5G
2,4,6-Trichlorophenyl formate ≥97.0% (by HPLC)
2,4,6-Trichlorophenyl formate

CAS Number: 4525-65-9
Molecular Formula: C7H3Cl3O2
Molecular Weight: 225.45
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 83


Product Categories: Synthetic Organic Chemistry, Carbon Monoxide (CO) Surrogate Reagents
Other Product Names / Synonyms: Formic Acid 2,4,6-Trichlorophenyl Ester

Formula: C₇H₃Cl₃O₂
MW: 225.45 g/mol
Melting Pt: 83 °C
MDL Number: MFCD27996252
CAS Number: 4525-65-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f8a1352773feca3e04b69a6f790a579c765f5419&vid=f8a1352773feca3e04b69a6f790a579c765f5419&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR