N,N,2-Trimethyl-5-nitrobenzenesulfonamide ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
T3112-1G
T3112-200MG
TCT3112-1GEA
193.39
CAD
TCT3112-1G
TCT3112-200MG
N,N,2-Trimethyl-5-nitrobenzenesulfonamide ≥98.0% (by HPLC, total nitrogen)
N,N,2-Trimethyl-5-nitrobenzenesulfonamide
CAS Number: 433695-36-4
MDL Number: MFCD03039912
Molecular Formula: C9H12N2O4S
Molecular Weight: 244.27
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 73
Storage Temperature: 0-10°C
Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Regenerative Medicine Research Reagents
Other Product Names / Synonyms: BRL 50481
Formula:
C₉H₁₂N₂O₄S Melting Pt: 73 °C |
MDL Number:
MFCD03039912 CAS Number: 433695-36-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...