3-Tetradecylthiophene ≥98.0%

Supplier: TCI America

T3104-25G T3104-5G
TCT3104-25GEA 476.05 CAD
TCT3104-25G TCT3104-5G
3-Tetradecylthiophene ≥98.0%
3-Tetradecylthiophene

CAS Number: 110851-66-6
MDL Number: MFCD09750675
Molecular Formula: C18H32S
Molecular Weight: 280.51
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 0.90
Lambda max.: 244 nm (CHCl3)

Formula: C₁₈H₃₂S
MW: 280.51 g/mol
Density: 0.90 (20 °C)
MDL Number: MFCD09750675
CAS Number: 110851-66-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9997405a4e66cc062441ddcf0e243c48eddba92&vid=a9997405a4e66cc062441ddcf0e243c48eddba92&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR