Talniflumate ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: 3-Oxo-1,3-dihydroisobenzofuran-1-yl 2-((3-(trifluoromethyl)phenyl)amino)nicotinate

T3101-1G T3101-200MG
TCT3101-1GEA 518.24 CAD
TCT3101-1G TCT3101-200MG
Talniflumate ≥98.0% (by HPLC, total nitrogen)
Talniflumate

CAS Number: 66898-62-2
MDL Number: MFCD00866135
Molecular Formula: C21H13F3N2O4
Molecular Weight: 414.34
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Color: Slightly Pale Yellow
Melting point (°C): 168
Lambda max.: 287 nm (CHCl3)Storage Temperature: 0-10°C

Formula: C₂₁H₁₃F₃N₂O₄
MW: 414.34 g/mol
Melting Pt: 168 °C
MDL Number: MFCD00866135
CAS Number: 66898-62-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d66860ef19a2b0fded445a8530f2ed7cfb55b91&vid=7d66860ef19a2b0fded445a8530f2ed7cfb55b91&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR