1-(p-Tolyl)-1-cyclohexanecarboxylic acid ≥98.0%

Supplier: TCI America

T3097-25G T3097-5G
TCT3097-25GEA 466.1 CAD
TCT3097-25G TCT3097-5G
1-(p-Tolyl)-1-cyclohexanecarboxylic acid ≥98.0%
1-(p-Tolyl)-1-cyclohexanecarboxylic acid

CAS Number: 84682-27-9
MDL Number: MFCD00065264
Molecular Formula: C14H18O2
Molecular Weight: 218.30
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 171

Formula: C₁₄H₁₈O₂
Melting Pt: 171 °C
MDL Number: MFCD00065264
CAS Number: 84682-27-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6dfcd91f3940af808e9c8d37fefcba04954dadaf&vid=6dfcd91f3940af808e9c8d37fefcba04954dadaf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR