3-[3-(Trifluoromethyl)phenyl]-1-propanol ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 3-(3-(Trifluoromethyl)phenyl)propan-1-ol

T3095-1G T3095-5G
TCT3095-1GEA 67.94 CAD
TCT3095-1G TCT3095-5G
3-[3-(Trifluoromethyl)phenyl]-1-propanol ≥98.0% (by GC)
3-[3-(Trifluoromethyl)phenyl]-1-propanol

CAS Number: 78573-45-2
MDL Number: MFCD08706408
Molecular Formula: C10H11F3O
Molecular Weight: 204.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 60
Specific Gravity (20/20): 1.21

Formula: C₁₀H₁₁F₃O
MW: 204.19 g/mol
Boiling Pt: 60 °C
Density: 1.21 (20 °C)
MDL Number: MFCD08706408
CAS Number: 78573-45-2
https://ca.vwr.com/stibo/web/std.lang.all/66/95/28996695.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR