4,4,5,5-Tetramethyl-2-(3-perylenyl)-1,3,2-dioxaborolane ≥97.0% (by GC)

Supplier: TCI America

T3089-1G T3089-200MG
TCT3089-1GEA 963.64 CAD
TCT3089-1G TCT3089-200MG
4,4,5,5-Tetramethyl-2-(3-perylenyl)-1,3,2-dioxaborolane ≥97.0% (by GC)
4,4,5,5-Tetramethyl-2-(3-perylenyl)-1,3,2-dioxaborolane

CAS Number: 950761-81-6
Molecular Formula: C26H23BO2
Molecular Weight: 378.28
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Color: Pale Yellow
Melting point (°C): 237
Lambda max.: 451 nm (Toluene)


Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives, Materials Chemistry, Building Blocks for Small Molecule Semiconductors
Other Product Names / Synonyms: (3-Perylenyl)boronic Acid Pinacol Ester, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)perylene

Formula: C₂₆H₂₃BO₂
Melting Pt: 237 °C
MDL Number: MFCD28098461
CAS Number: 950761-81-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=21cd9c0295bcadbf4904b8ec96ace32e048ab5ba&vid=21cd9c0295bcadbf4904b8ec96ace32e048ab5ba&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR