1,3,5-Tris(4-iodophenyl)benzene ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 4,4''-Diiodo-5'-(4-iodophenyl)-1,1':3',1''-terphenyl

T3084-1G T3084-5G
TCT3084-1GEA 211.72 CAD
TCT3084-1G TCT3084-5G
1,3,5-Tris(4-iodophenyl)benzene ≥98.0% (by HPLC)
1,3,5-Tris(4-iodophenyl)benzene

CAS Number: 151417-38-8
MDL Number: MFCD07369795
Molecular Formula: C24H15I3
Molecular Weight: 684.10
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 266

Formula: C₂₄H₁₅I₃
MW: 684.10 g/mol
Melting Pt: 266 °C
MDL Number: MFCD07369795
CAS Number: 151417-38-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=79451e8ae7c6da4830908c15a92a7996a0be142c&vid=79451e8ae7c6da4830908c15a92a7996a0be142c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR