1-(4-(Trifluoromethyl)benzyl)piperazine ≥97.0%

Supplier: TCI America

Synonyms: 1-(4-Trifluoromethylbenzyl)piperazine

T3075-1G T3075-5G
TCT3075-1GEA 85.95 CAD
TCT3075-1G TCT3075-5G
1-(4-(Trifluoromethyl)benzyl)piperazine ≥97.0%
1-(4-(Trifluoromethyl)benzyl)piperazine

CAS Number: 107890-32-4
MDL Number: MFCD03407490
Molecular Formula: C12H15F3N2
Molecular Weight: 244.26
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Color: Slightly Pale Yellow
Boiling point (°C): 100
Flash Point (°C): 110
Specific Gravity (20/20): 1.18

Formula: C₁₂H₁₅F₃N₂
Boiling Pt: 100 °C
Density: 1.18 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD03407490
CAS Number: 107890-32-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e69df65f46e4373069ab9b0b317c8fb30cab5055&vid=e69df65f46e4373069ab9b0b317c8fb30cab5055&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR