1,3,5-Tris(3-bromophenyl)benzene ≥96.0% (by HPLC)

Supplier: TCI America

Synonyms: 3,3''-Dibromo-5'-(3-bromophenyl)-1,1':3',1''-terphenyl

T3073-1G T3073-5G
TCT3073-1GEA 97.96 CAD
TCT3073-1G TCT3073-5G
1,3,5-Tris(3-bromophenyl)benzene ≥96.0% (by HPLC)
1,3,5-Tris(3-bromophenyl)benzene

CAS Number: 96761-85-2
MDL Number: MFCD02323390
Molecular Formula: C24H15Br3
Molecular Weight: 543.10
Purity/Analysis Method: >96.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 172

Formula: C₂₄H₁₅Br₃
MW: 543.10 g/mol
Melting Pt: 172 °C
MDL Number: MFCD02323390
CAS Number: 96761-85-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c335c0f1f1fe392fb92461551969bae6a97528f&vid=8c335c0f1f1fe392fb92461551969bae6a97528f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR