Triethyl orthobenzoate ≥97.0% (by GC)
Supplier: TCI America
T3070-25G
T3070-5G
TCT3070-25GEA
205.75
CAD
TCT3070-25G
TCT3070-5G
Triethyl orthobenzoate ≥97.0% (by GC)
Triethyl orthobenzoate
CAS Number: 1663-61-2
MDL Number: MFCD00009222
Molecular Formula: C13H20O3
Molecular Weight: 224.30
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 240
Flash Point (°C): 97
Specific Gravity (20/20): 0.99
Other Product Names / Synonyms: Orthobenzoic Acid Triethyl Ester, alpha,alpha,alpha-Triethoxytoluene
Formula:
C₁₃H₂₀O₃ Boiling Pt: 240 °C Density: 0.99 (20 °C) Flash Pt: 97 °C |
MDL Number:
MFCD00009222 CAS Number: 1663-61-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...