2-(2-Thienyl)ethyl-p-toluenesulfonate ≥95.0%
Supplier: TCI America
Synonyms:
p-Toluenesulfonic acid 2-(2-Thienyl)ethyl Ester, 2-(Thiophen-2-yl)ethyl 4-methylbenzenesulphonate, p-Toluenesulphonic acid 2-(2-Thienyl)ethyl Ester, 2-(Thiophen-2-yl)ethyl 4-methylbenzenesulfonate
CAS Number: 40412-06-4
MDL Number: MFCD05150366
Molecular Formula: C13H14O3S2
Molecular Weight: 282.37
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Color: Pale Yellow
Melting point (°C): 32
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives
Other Product Names / Synonyms: p-Toluenesulfonic Acid 2-(2-Thienyl)ethyl Ester
Formula:
C₁₃H₁₄O₃S₂ MW: 282.37 g/mol Melting Pt: 32 °C |
MDL Number:
MFCD05150366 CAS Number: 40412-06-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...