Tolnaftate ≥97.0%

Supplier: TCI America

Synonyms: O-2-Naphthyl Methyl(3-methylphenyl)carbamothioate, O-Naphthalen-2-yl methyl(m-tolyl)carbamothioate, Methyl(3-methylphenyl)carbamothioic Acid O-2-Naphthyl Ester

T3059-1G T3059-5G
TCT3059-1GEA 75.84 CAD
TCT3059-1G TCT3059-5G
Tolnaftate ≥97.0%
Tolnaftate

CAS Number: 2398-96-1
MDL Number: MFCD00056611
Molecular Formula: C19H17NOS
Molecular Weight: 307.41
Purity/Analysis Method: >97.0% (HPLC,N)
Form: Crystal
Color: White
Melting point (°C): 113
Lambda max.: 258 nm (MeOH)Storage Temperature: 0-10°C


Product Categories: Bioscience, Antifungal Ingredients
Other Product Names / Synonyms: O-2-Naphthyl Methyl(3-methylphenyl)carbamothioate, Methyl(3-methylphenyl)carbamothioic Acid O-2-Naphthyl Ester

Formula: C₁₉H₁₇NOS
MW: 307.41 g/mol
Melting Pt: 113 °C
MDL Number: MFCD00056611
CAS Number: 2398-96-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1b5ea911f41566f0ecccdbb3ce486793225ece7&vid=e1b5ea911f41566f0ecccdbb3ce486793225ece7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR