3,3',5,5'-Tetra-tert-butyl-2,2'-dihydroxybiphenyl ≥98.0% (by GC)

Supplier: TCI America

T3052-1G T3052-5G
TCT3052-1GEA 112.18 CAD
TCT3052-1G TCT3052-5G
3,3',5,5'-Tetra-tert-butyl-2,2'-dihydroxybiphenyl ≥98.0% (by GC)
3,3',5,5'-Tetra-tert-butyl-2,2'-dihydroxybiphenyl

CAS Number: 6390-69-8
MDL Number: MFCD00085287
Molecular Formula: C28H42O2
Molecular Weight: 410.64
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 200
Lambda max.: 283 nm


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 3,3',5,5'-Tetra-tert-butylbiphenyl-2,2'-diol

Formula: C₂₈H₄₂O₂
Melting Pt: 200 °C
MDL Number: MFCD00085287
CAS Number: 6390-69-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a317434e0e284dec1c365cfa8ff31ff4eaaea86d&vid=a317434e0e284dec1c365cfa8ff31ff4eaaea86d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR