1,3,6,8-Tetraphenylpyrene ≥98.0% (by HPLC)
Supplier: TCI America
T3042-200MG
T3042-50MG
TCT3042-200MGEA
432.92
CAD
TCT3042-200MG
TCT3042-50MG
1,3,6,8-Tetraphenylpyrene ≥98.0% (by HPLC)
1,3,6,8-Tetraphenylpyrene
CAS Number: 13638-82-9
MDL Number: MFCD00021662
Molecular Formula: C40H26
Molecular Weight: 506.65
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: Pale Yellow
Melting point (°C): 299
Lambda max.: 478 nm (CH2Cl2)
Formula:
C₄₀H₂₆ Melting Pt: 299 °C |
MDL Number:
MFCD00021662 CAS Number: 13638-82-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...