Tris(1,1,1,3,3,3-hexafluoro-2-propyl)phosphate ≥98.0%
Supplier: TCI America
Synonyms:
Phosphoric Acid Tris(1,1,1,3,3,3-hexafluoro-2-propyl) Ester
T3041-1G
T3041-5G
TCT3041-1GEA
224.36
CAD
TCT3041-1G
TCT3041-5G
Tris(1,1,1,3,3,3-hexafluoro-2-propyl)phosphate ≥98.0%
Tris(1,1,1,3,3,3-hexafluoro-2-propyl)phosphate
CAS Number: 66489-68-7
Molecular Formula: C9H3F18O4P
Molecular Weight: 548.06
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 72
Melting point (°C): 32
Storage Temperature: 0-10°C
Product Categories: Materials Chemistry, Secondary Battery Research Reagents
Other Product Names / Synonyms: Phosphoric Acid Tris(1,1,1,3,3,3-hexafluoro-2-propyl) Ester
Formula:
C₉H₃F₁₈O₄P MW: 548.06 g/mol Boiling Pt: 72 °C Melting Pt: 32 °C |
CAS Number:
66489-68-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...