3-(Trimethylsilyl)propiolic acid ≥97.0% (by GC, titration analysis)

Supplier: TCI America

T3040-1G T3040-5G
TCT3040-1GEA 592.5 CAD
TCT3040-1G TCT3040-5G
3-(Trimethylsilyl)propiolic acid ≥97.0% (by GC, titration analysis)
3-(Trimethylsilyl)propiolic acid

CAS Number: 5683-31-8
MDL Number: MFCD00190216
Molecular Formula: C6H10O2Si
Molecular Weight: 142.23
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 110
Melting point (°C): 48
Flash Point (°C): 96
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 3-(Trimethylsilyl)propargylic Acid

Formula: C₆H₁₀O₂Si
Boiling Pt: 110 °C
Melting Pt: 48 °C
Flash Pt: 96 °C
MDL Number: MFCD00190216
CAS Number: 5683-31-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a76e59dd450de8445f3ba76d18e8415fe45a617f&vid=a76e59dd450de8445f3ba76d18e8415fe45a617f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR