1-[(Triisopropylsilyl)ethynyl]-1,2-benziodoxol-3(1H)-one ≥98.0%

Supplier: TCI America

Synonyms: TIPS-EBX, 1-((Tris(1-methylethyl)silyl)ethynyl)-1,2-benziodoxol-3(1H)-one

T3039-1G T3039-200MG
TCT3039-1GEA 320.74 CAD
TCT3039-1G TCT3039-200MG
1-[(Triisopropylsilyl)ethynyl]-1,2-benziodoxol-3(1H)-one ≥98.0%
1-[(Triisopropylsilyl)ethynyl]-1,2-benziodoxol-3(1H)-one

CAS Number: 181934-30-5
MDL Number: MFCD18632570
Molecular Formula: C18H25IO2Si
Molecular Weight: 428.39
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 170


Product Categories: Synthetic Organic Chemistry, Hypervalent Iodine Compounds, Silicon Compounds (for Synthesis), Click Chemistry
Other Product Names / Synonyms: TIPS-EBX

Formula: C₁₈H₂₅IO₂Si
MW: 428.39 g/mol
Melting Pt: 170 °C
MDL Number: MFCD18632570
CAS Number: 181934-30-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=12135aee5b939cd0981af53cc89f80f7063a9af6&vid=12135aee5b939cd0981af53cc89f80f7063a9af6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR