Trimethyl-1,3,5-cyclohexanetricarboxylate (cis and trans mixture) ≥98.0%
Supplier: TCI America
Synonyms:
1,3,5-Cyclohexanetricarboxylic Acid Trimethyl Ester
T3033-1G
T3033-5G
TCT3033-1GEA
323.9
CAD
TCT3033-1G
TCT3033-5G
Trimethyl-1,3,5-cyclohexanetricarboxylate (cis and trans mixture) ≥98.0%
Trimethyl-1,3,5-cyclohexanetricarboxylate (cis and trans mixture)
CAS Number: 52831-11-5
Molecular Formula: C12H18O6
Molecular Weight: 258.27
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 46
Other Product Names / Synonyms: 1,3,5-Cyclohexanetricarboxylic Acid Trimethyl Ester
Formula:
C₁₂H₁₈O₆ MW: 258.27 g/mol Melting Pt: 46 °C |
CAS Number:
52831-11-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...