4'-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)biphenyl-4-ol ≥98.0% (by GC)

Supplier: TCI America

T3031-1G T3031-5G
TCT3031-1GEA 206.98 CAD
TCT3031-1G TCT3031-5G
4'-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)biphenyl-4-ol ≥98.0% (by GC)
4'-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)biphenyl-4-ol

CAS Number: 760989-91-1
MDL Number: MFCD22414472
Molecular Formula: C18H21BO3
Molecular Weight: 296.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 150


Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives
Other Product Names / Synonyms: 2-(4'-Hydroxybiphenyl-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, (4'-Hydroxybiphenyl-4-yl)boronic Acid Pinacol Ester

Formula: C₁₈H₂₁BO₃
Melting Pt: 150 °C
MDL Number: MFCD22414472
CAS Number: 760989-91-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=01dd377d91b021adb46c578c8a37ea50004d8348&vid=01dd377d91b021adb46c578c8a37ea50004d8348&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR