Trimethyl-1,2,4-cyclohexanetricarboxylate (cis and trans mixture) ≥97.0% (by GC)
Supplier: TCI America
T3029-1G
T3029-5G
TCT3029-1GEA
219.62
CAD
TCT3029-1G
TCT3029-5G
Trimethyl-1,2,4-cyclohexanetricarboxylate (cis and trans mixture) ≥97.0% (by GC)
Trimethyl-1,2,4-cyclohexanetricarboxylate (cis and trans mixture)
CAS Number: 185855-30-5
Molecular Formula: C12H18O6
Molecular Weight: 258.27
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 1.20
Storage Temperature: 0-10°C
Other Product Names / Synonyms: 1,2,4-Cyclohexanetricarboxylic Acid Trimethyl Ester
Formula:
C₁₂H₁₈O₆ Density: 1.20 (20 °C) |
CAS Number:
185855-30-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...