4-(1,1,2,2-Tetrafluoroethoxy)benzoic acid ≥98.0%
Supplier: TCI America
T3027-25G
T3027-5G
TCT3027-25GEA
312.84
CAD
TCT3027-25G
TCT3027-5G
4-(1,1,2,2-Tetrafluoroethoxy)benzoic acid ≥98.0%
4-(1,1,2,2-Tetrafluoroethoxy)benzoic acid
CAS Number: 10009-25-3
MDL Number: MFCD00155926
Molecular Formula: C9H6F4O3
Molecular Weight: 238.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 178
Formula:
C₉H₆F₄O₃ Melting Pt: 178 °C |
MDL Number:
MFCD00155926 CAS Number: 10009-25-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...