4-(1H-1,2,4-Triazol-1-yl)aniline ≥98.0%
Supplier: TCI America
Synonyms:
4-(1,2,4-Triazol-1-yl)aniline, 1-(4-Aminophenyl)-1,2,4-triazole
T3013-1G
T3013-200MG
TCT3013-1GEA
426.6
CAD
TCT3013-1G
TCT3013-200MG
4-(1H-1,2,4-Triazol-1-yl)aniline ≥98.0%
4-(1H-1,2,4-Triazol-1-yl)aniline
CAS Number: 6523-49-5
MDL Number: MFCD00171470
Molecular Formula: C8H8N4
Molecular Weight: 160.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 143
Other Product Names / Synonyms: 1-(4-Aminophenyl)-1,2,4-triazole
Formula:
C₈H₈N₄ MW: 160.1759 g/mol Melting Pt: 143 °C |
MDL Number:
MFCD00171470 CAS Number: 6523-49-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...