3-(Trifluoromethyl)-5,6,7,8-tetrahydro[1,2,4]triazolo[4,3-a]pyrazine hydrochloride ≥98.0%
Supplier: TCI America
Synonyms:
3-(Trifluoromethyl)-5H,6H,7H,8H-[1,2,4]triazolo[4,3-a]pyrazine hydrochloride, 3-(Trifluoromethyl)-5,6,7,8-tetrahydro-[1,2,4]triazolo[4,3-a]pyrazine hydrochloride, 3-Trifluoromethyl-5,6,7,8-tetrahydro-[1,2,4]triazolo[4,3-a]pyrazine hydrochloride
T2998-1G
T2998-5G
TCT2998-1GEA
86.9
CAD
TCT2998-1G
TCT2998-5G
3-(Trifluoromethyl)-5,6,7,8-tetrahydro[1,2,4]triazolo[4,3-a]pyrazine hydrochloride ≥98.0%
3-(Trifluoromethyl)-5,6,7,8-tetrahydro[1,2,4]triazolo[4,3-a]pyrazine hydrochloride
CAS Number: 762240-92-6
MDL Number: MFCD09817638
Molecular Formula: C6H7F3N4
Molecular Weight: 228.60
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 264
Formula:
C6H8ClF3N4 MW: 228.60 g/mol Melting Pt: 264 °C |
MDL Number:
MFCD09817638 CAS Number: 762240-92-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...