4,4,4-Trifluoro-1-(p-tolyl)-1,3-butanedione ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 4,4,4-Trifluoro-1-(p-tolyl)butane-1,3-dione, 4,4,4-Trifluoro-1-(4-methylphenyl)-1,3-butanedione

T2997-25G T2997-5G
TCT2997-25GEA 191.18 CAD
TCT2997-25G TCT2997-5G
4,4,4-Trifluoro-1-(p-tolyl)-1,3-butanedione ≥98.0% (by GC)
4,4,4-Trifluoro-1-(p-tolyl)-1,3-butanedione

CAS Number: 720-94-5
MDL Number: MFCD00517909
Molecular Formula: C11H9F3O2
Molecular Weight: 230.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 47


Product Categories: Synthetic Organic Chemistry, Ligands, Fluorinated Building Blocks
Other Product Names / Synonyms: 4,4,4-Trifluoro-1-(4-methylphenyl)-1,3-butanedione

Formula: C₁₁H₉F₃O₂
MW: 230.19 g/mol
Melting Pt: 47 °C
MDL Number: MFCD00517909
CAS Number: 720-94-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b9390c9b12b3e2afa7c5898ff7d29cc821b24bd&vid=3b9390c9b12b3e2afa7c5898ff7d29cc821b24bd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR