Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine ≥97.0%

Supplier: TCI America

Synonyms: TBTA, Tris((1-benzyl-1H-1,2,3-triazol-4-yl)methyl)amine

T2993-1G T2993-5G
TCT2993-1GEA 357.85 CAD
TCT2993-1G TCT2993-5G
Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine ≥97.0%
Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine

CAS Number: 510758-28-8
MDL Number: MFCD09265124
Molecular Formula: C30H30N10
Molecular Weight: 530.64
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 138
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Ligands, Metal Catalysts & Ligands (Click Chemistry)
Other Product Names / Synonyms: TBTA

Formula: C₃₀H₃₀N₁₀
MW: 530.64 g/mol
Melting Pt: 138 °C
MDL Number: MFCD09265124
CAS Number: 510758-28-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=01e6c7b918e1e48e634021bc5a9d26238abc7072&vid=01e6c7b918e1e48e634021bc5a9d26238abc7072&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR