Tanshinone IIA ≥97.0%

Supplier: TCI America

Synonyms: Dan Shen Ketone, 1,6,6-Trimethyl-6,7,8,9-tetrahydrophenanthro[1,2-b]furan-10,11-dione

T2987-10MG T2987-50MG
TCT2987-10MGEA 216.46 CAD
TCT2987-10MG TCT2987-50MG
Tanshinone IIA ≥97.0%
Tanshinone IIA

CAS Number: 568-72-9
MDL Number: MFCD00238692
Molecular Formula: C19H18O3
Molecular Weight: 294.35
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Color: Reddish Yellow
Melting point (°C): 198
Storage Temperature: 0-10°C


Product Categories: Bioscience, Others (Phytochemicals)
Other Product Names / Synonyms: Dan Shen Ketone, 1,6,6-Trimethyl-6,7,8,9-tetrahydrophenanthro[1,2-b]furan-10,11-dione

Formula: C₁₉H₁₈O₃
Melting Pt: 198 °C
MDL Number: MFCD00238692
CAS Number: 568-72-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c3926a6d2b29bd82b0984e3d19c5fae695d9f809&vid=c3926a6d2b29bd82b0984e3d19c5fae695d9f809&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR