Tiropramide hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

T2978-1G T2978-5G
TCT2978-1GEA 186.44 CAD
TCT2978-1G TCT2978-5G
Tiropramide hydrochloride ≥98.0% (by HPLC, titration analysis)
Tiropramide hydrochloride

CAS Number: 53567-47-8
MDL Number: MFCD01741847
Molecular Formula: C28H41N3O3
Molecular Weight: 504.11
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 182

Formula: C₂₈H₄₁N₃O₃
MW: 504.11 g/mol
Melting Pt: 182 °C
MDL Number: MFCD01741847
CAS Number: 53567-47-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=018796ae01fa41213531e9f5fd4221926ebed0c1&vid=018796ae01fa41213531e9f5fd4221926ebed0c1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR