5-(Trifluoromethyl)picolinic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

T2975-1G T2975-5G
TCT2975-1GEA 55.3 CAD
TCT2975-1G TCT2975-5G
5-(Trifluoromethyl)picolinic acid ≥98.0% (by GC, titration analysis)
5-(Trifluoromethyl)picolinic acid

CAS Number: 80194-69-0
MDL Number: MFCD04113632
Molecular Formula: C7H4F3NO2
Molecular Weight: 191.11
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 137


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 5-(Trifluoromethyl)picolinic Acid

Formula: C₇H₄F₃NO₂
Melting Pt: 137 °C
MDL Number: MFCD04113632
CAS Number: 80194-69-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fb05de297f5a2499e809dfc87c063166571600e8&vid=fb05de297f5a2499e809dfc87c063166571600e8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR