tri-Magnesium dicitrate nonahydrate ≥98.0%

Supplier: TCI America

Synonyms: Trimagnesium Dicitrate Nonahydrate, Magnesium 2-hydroxypropane-1,2,3-tricarboxylate nonahydrate, Magnesium citrate tribasic nonahydrate

T2966-25G T2966-500G
TCT2966-25GEA 41.08 CAD
TCT2966-25G TCT2966-500G
tri-Magnesium dicitrate nonahydrate ≥98.0%
tri-Magnesium dicitrate nonahydrate

CAS Number: 153531-96-5
MDL Number: MFCD00036422
Molecular Formula: C6H8O7
Molecular Weight: 451.11
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White

Formula: C₁₂H₁₀Mg₃O₁₄·9H₂O
MDL Number: MFCD00036422
CAS Number: 153531-96-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dec3ae5ba35cace20e5b12fe1c7934b7975195d5&vid=dec3ae5ba35cace20e5b12fe1c7934b7975195d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR