Tetrakis(4-bromophenyl)methane ≥95.0% (by GC)

Supplier: TCI America

T2960-1G T2960-5G
TCT2960-1GEA 284.4 CAD
TCT2960-1G TCT2960-5G
Tetrakis(4-bromophenyl)methane ≥95.0% (by GC)
Tetrakis(4-bromophenyl)methane

CAS Number: 105309-59-9
MDL Number: MFCD19688472
Molecular Formula: C25H16Br4
Molecular Weight: 636.02
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Color: Slightly Pale Yellow
Melting point (°C): 300

Formula: C₂₅H₁₆Br₄
Melting Pt: 300 °C
MDL Number: MFCD19688472
CAS Number: 105309-59-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ceff15eae8e0465e19557a41dc9b356487d896b&vid=8ceff15eae8e0465e19557a41dc9b356487d896b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR